Assay Details
| Gene Symbol: mxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298114.1 | NP_001285043.1 | 5 - 6 | 4927 | 62 | |
| NM_132320.3 | NP_572548.1 | 5 - 6 | 4927 | 62 | ||
More Information
Set Membership: |
|

mxc
Genomic Map