Assay Details
| Gene Symbol: nej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201646.2 | NP_001188575.1 | 2 | 1189 | 131 | |
| NM_001201647.2 | NP_001188576.1 | 2 | 1159 | 131 | ||
| NM_001272458.1 | NP_001259387.1 | 2 | 1091 | 131 | ||
| NM_079903.4 | NP_524642.2 | 2 | 1236 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001201646.2, NM_001201647.2, NM_001272458.1, NM_079903.4 transcripts.
Set Membership: |
|

nej
Genomic Map