Assay Details
Gene Symbol: Ser7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132354.3 | NP_572582.1 | 1 - 2 | 160 | 76 | |
GenBank mRNA | AY051477.1 | - | 1 - 2 | 151 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
