Assay Details
| Gene Symbol: Ser7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132354.3 | NP_572582.1 | 2 - 3 | 311 | 90 | |
| GenBank mRNA | AY051477.1 | - | 2 - 3 | 302 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ser7| Gene Symbol: Ser7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132354.3 | NP_572582.1 | 2 - 3 | 311 | 90 | |
| GenBank mRNA | AY051477.1 | - | 2 - 3 | 302 | 90 | |
Set Membership: |
|