Assay Details
| Gene Symbol: RhoU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042801.2 | NP_001036266.1 | 4 - 5 | 1518 | 73 | |
| NM_001042802.2 | NP_001036267.1 | 4 - 5 | 1518 | 73 | ||
| NM_001298144.1 | NP_001285073.1 | 3 - 4 | 1480 | 73 | ||
| GenBank mRNA | AY075502.1 | - | 4 - 5 | 1511 | 73 | |
More Information
Set Membership: |
|

RhoU
Genomic Map