Assay Details
| Gene Symbol: RhoU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042801.2 | NP_001036266.1 | 7 - 8 | 2059 | 68 | |
| NM_001298144.1 | NP_001285073.1 | 6 - 7 | 2021 | 68 | ||
More Information
Set Membership: |
|

RhoU
Genomic Map