Assay Details
| Gene Symbol: ATPsyndelta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272468.1 | NP_001259397.1 | 2 - 3 | 306 | 82 | |
| NM_001298145.1 | NP_001285074.1 | 2 - 3 | 306 | 82 | ||
| NM_132382.3 | NP_572610.1 | 2 - 3 | 306 | 82 | ||
| GenBank mRNA | AY071203.1 | - | 2 - 3 | 304 | 82 | |
More Information
Set Membership: |
|

ATPsyndelta
Genomic Map