Assay Details
Gene Symbol: CG2186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272504.1 | NP_001259433.1 | 4 - 5 | 1262 | 57 | |
NM_132432.2 | NP_572660.1 | 4 - 5 | 1262 | 57 |

More Information
Set Membership: |
![]() ![]() |
