Assay Details
| Gene Symbol: Pa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132484.4 | NP_572712.1 | 1 - 2 | 303 | 58 | |
| GenBank mRNA | AY070686.1 | - | 1 - 2 | 302 | 58 | |
| AY071007.1 | - | 1 - 2 | 301 | 58 | ||
| KX532091.1 | - | 1 - 2 | 210 | 58 | ||
More Information
Set Membership: |
|

Pa1
Genomic Map