Assay Details
| Gene Symbol: Sk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132489.2 | NP_572717.1 | 2 | 2530 | 79 | |
| NM_167286.2 | NP_727528.1 | 2 | 2464 | 79 | ||
| GenBank mRNA | AY119652.1 | - | 2 | 2462 | 79 | |
More Information
Set Membership: |
|

Sk1
Genomic Map