Assay Details
| Gene Symbol: CG43155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132492.3 | NP_572720.2 | 2 - 3 | 502 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG43155| Gene Symbol: CG43155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132492.3 | NP_572720.2 | 2 - 3 | 502 | 93 | |
Set Membership: |
|