Assay Details
| Gene Symbol: Uba5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132494.3 | NP_572722.2 | 1 - 2 | 418 | 58 | |
| GenBank mRNA | BT150207.1 | - | 1 - 2 | 429 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uba5| Gene Symbol: Uba5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132494.3 | NP_572722.2 | 1 - 2 | 418 | 58 | |
| GenBank mRNA | BT150207.1 | - | 1 - 2 | 429 | 58 | |
Set Membership: |
|