Assay Details
| Gene Symbol: CG2247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258696.2 | NP_001245625.1 | 4 - 5 | 1008 | 69 | |
| NM_001258698.2 | NP_001245627.1 | 4 - 5 | 1008 | 69 | ||
| NM_132507.2 | NP_572735.1 | 4 - 5 | 1008 | 69 | ||
| GenBank mRNA | BT031131.1 | - | 4 - 5 | 934 | 69 | |
More Information
Set Membership: |
|

CG2247
Genomic Map