Assay Details
| Gene Symbol: RPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298202.1 | NP_001285131.1 | 1 - 2 | 324 | 70 | |
| NM_132509.2 | NP_572737.1 | 1 - 2 | 324 | 70 | ||
| GenBank mRNA | AY113602.1 | - | 1 - 2 | 325 | 70 | |
| KX532026.1 | - | 1 - 2 | 238 | 70 | ||
More Information
Set Membership: |
|

RPA3
Genomic Map