Assay Details
| Gene Symbol: wisp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298215.1 | NP_001285144.1 | 2 - 3 | 1812 | 60 | |
| NM_132538.2 | NP_572766.1 | 3 - 4 | 1619 | 60 | ||
| GenBank mRNA | BT149893.1 | - | 2 - 3 | 1810 | 60 | |
More Information
Set Membership: |
|

wisp
Genomic Map