Assay Details
| Gene Symbol: wisp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298214.1 | NP_001285143.1 | 5 - 6 | 3560 | 63 | |
| NM_001298215.1 | NP_001285144.1 | 4 - 5 | 3756 | 63 | ||
| NM_132538.2 | NP_572766.1 | 5 - 6 | 3563 | 63 | ||
| GenBank mRNA | BT149893.1 | - | 4 - 5 | 3754 | 63 | |
More Information
Set Membership: |
|

wisp
Genomic Map