Assay Details
| Gene Symbol: wisp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298214.1 | NP_001285143.1 | 7 - 8 | 3922 | 117 | |
| NM_001298215.1 | NP_001285144.1 | 6 - 7 | 4118 | 117 | ||
| NM_132538.2 | NP_572766.1 | 7 - 8 | 3925 | 117 | ||
| GenBank mRNA | BT149893.1 | - | 6 - 7 | 4116 | 117 | |
More Information
Set Membership: |
|

wisp
Genomic Map