Assay Details
| Gene Symbol: Uch-L5R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132553.1 | NP_572781.1 | 1 | 1126 | 112 | |
| GenBank mRNA | BT150419.1 | - | 1 | 1126 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uch-L5R| Gene Symbol: Uch-L5R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132553.1 | NP_572781.1 | 1 | 1126 | 112 | |
| GenBank mRNA | BT150419.1 | - | 1 | 1126 | 112 | |
Set Membership: |
|