Assay Details
| Gene Symbol: Cpr11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132575.3 | NP_572803.1 | 2 - 3 | 340 | 61 | |
| GenBank mRNA | AY071531.1 | - | 2 - 3 | 337 | 61 | |
More Information
Set Membership: |
|

Cpr11A
Genomic Map