Assay Details
| Gene Symbol: Cpr11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132579.3 | NP_572807.1 | 2 - 3 | 595 | 71 | |
| GenBank mRNA | AY060775.1 | - | 2 - 3 | 578 | 71 | |
More Information
Set Membership: |
|

Cpr11B
Genomic Map