Assay Details
| Gene Symbol: Brms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132612.2 | NP_572840.1 | 1 - 2 | 374 | 61 | |
| GenBank mRNA | AY102677.1 | - | 1 - 2 | 351 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brms1| Gene Symbol: Brms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132612.2 | NP_572840.1 | 1 - 2 | 374 | 61 | |
| GenBank mRNA | AY102677.1 | - | 1 - 2 | 351 | 61 | |
Set Membership: |
|