Assay Details
| Gene Symbol: Tim9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298279.1 | NP_001285208.1 | 1 - 2 | 138 | 73 | |
| NM_132653.2 | NP_572881.1 | 1 - 2 | 193 | 73 | ||
| GenBank mRNA | BT025812.1 | - | 1 - 2 | 111 | 73 | |
| KX531321.1 | - | 1 - 2 | 66 | 73 | ||
More Information
Set Membership: |
|

Tim9a
Genomic Map