Assay Details
| Gene Symbol: CtsB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272607.2 | NP_001259536.2 | 3 - 4 | 791 | 78 | |
| NM_132692.3 | NP_572920.1 | 3 - 4 | 791 | 78 | ||
| GenBank mRNA | AY060640.1 | - | 3 - 4 | 700 | 78 | |
More Information
Set Membership: |
|

CtsB1
Genomic Map