Assay Details
| Gene Symbol: Scamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272643.1 | NP_001259572.1 | 4 - 5 | 743 | 54 | |
| NM_132795.4 | NP_573023.2 | 4 - 5 | 743 | 54 | ||
| NM_167442.2 | NP_727850.1 | 4 - 5 | 1165 | 54 | ||
| GenBank mRNA | AF241834.1 | - | 4 - 5 | 636 | 54 | |
| AY075562.1 | - | 4 - 5 | 750 | 54 | ||
More Information
Set Membership: |
|

Scamp
Genomic Map