Assay Details
| Gene Symbol: shtd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132797.3 | NP_573025.2 | 5 - 6 | 4705 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

shtd| Gene Symbol: shtd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132797.3 | NP_573025.2 | 5 - 6 | 4705 | 62 | |
Set Membership: |
|