Assay Details
| Gene Symbol: CCT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132838.3 | NP_573066.1 | 1 - 2 | 260 | 88 | |
| GenBank mRNA | AY051470.1 | - | 1 - 2 | 239 | 88 | |
| Gene Symbol: Hsp70Bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT011379.1 | - | 2675 | 88 | ||
More Information
Set Membership: |
|

CCT6
Genomic Map