Assay Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298366.1 | NP_001285295.1 | 1 - 2 | 140 | 61 | |
| NM_132840.5 | NP_573068.3 | 1 - 2 | 140 | 61 | ||
| GenBank mRNA | AY069410.1 | - | 1 - 2 | 100 | 61 | |
More Information
Set Membership: |
|

Mvd
Genomic Map