Assay Details
| Gene Symbol: SWIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132888.2 | NP_573116.2 | 4 - 5 | 1935 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SWIP| Gene Symbol: SWIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132888.2 | NP_573116.2 | 4 - 5 | 1935 | 82 | |
Set Membership: |
|