Assay Details
| Gene Symbol: TH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298397.1 | NP_001285326.1 | 4 - 5 | 1788 | 97 | |
| NM_132895.3 | NP_573123.1 | 4 - 5 | 1788 | 97 | ||
| GenBank mRNA | AJ238377.1 | - | 4 - 5 | 1757 | 97 | |
| AJ238378.1 | - | 4 - 5 | 1754 | 97 | ||
| AY051957.1 | - | 4 - 5 | 1760 | 97 | ||
More Information
Set Membership: |
|

TH1
Genomic Map