Assay Details
| Gene Symbol: Nup153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103532.3 | NP_001097002.1 | 8 - 9 | 6532 | 76 | |
| NM_001144734.2 | NP_001138206.1 | 6 - 7 | 6478 | 76 | ||
| NM_132908.5 | NP_573136.3 | 7 - 8 | 6394 | 76 | ||
| GenBank mRNA | BT025178.1 | - | 6 - 7 | 6396 | 76 | |
More Information
Set Membership: |
|

Nup153
Genomic Map