Assay Details
| Gene Symbol: CG8945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132962.2 | NP_573190.1 | 2 - 3 | 3126 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG8945| Gene Symbol: CG8945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132962.2 | NP_573190.1 | 2 - 3 | 3126 | 73 | |
Set Membership: |
|