Assay Details
| Gene Symbol: TTLL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132969.3 | NP_573197.2 | 3 - 4 | 1338 | 61 | |
| GenBank mRNA | BT044430.1 | - | 3 - 4 | 1338 | 61 | |
More Information
Set Membership: |
|

TTLL1A
Genomic Map