Assay Details
| Gene Symbol: p-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132988.3 | NP_573216.1 | 3 - 4 | 1240 | 71 | |
| GenBank mRNA | AY075163.1 | - | 3 - 4 | 1240 | 71 | |
More Information
Set Membership: |
|

p-cup
Genomic Map