Assay Details
| Gene Symbol: p-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132988.3 | NP_573216.1 | 4 - 5 | 1386 | 91 | |
| GenBank mRNA | AY075163.1 | - | 4 - 5 | 1386 | 91 | |
More Information
Set Membership: |
|

p-cup
Genomic Map