Assay Details
| Gene Symbol: Mul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316436.1 | NP_001303365.1 | 1 - 2 | 184 | 71 | |
| NM_139590.4 | NP_647847.1 | 1 - 2 | 184 | 71 | ||
| GenBank mRNA | AY113263.1 | - | 1 - 2 | 200 | 71 | |
More Information
Set Membership: |
|

Mul1
Genomic Map