Assay Details
| Gene Symbol: Rcd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139595.3 | NP_647852.1 | 1 - 2 | 377 | 101 | |
| GenBank mRNA | AY060599.1 | - | 1 - 2 | 332 | 101 | |
More Information
Set Membership: |
|

Rcd5
Genomic Map