Assay Details
| Gene Symbol: CG43367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202119.2 | NP_001189048.2 | 12 - 13 | 4514 | 68 | |
| NM_001259684.1 | NP_001246613.1 | 11 - 12 | 4716 | 68 | ||
| NM_001259685.1 | NP_001246614.1 | 10 - 11 | 4363 | 68 | ||
| NM_139612.3 | NP_647869.3 | 11 - 12 | 4388 | 68 | ||
More Information
Set Membership: |
|

CG43367
Genomic Map