Assay Details
| Gene Symbol: CG43367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202119.2 | NP_001189048.2 | 3 - 4 | 890 | 67 | |
| NM_001259684.1 | NP_001246613.1 | 3 - 4 | 890 | 67 | ||
| NM_001259685.1 | NP_001246614.1 | 2 - 3 | 537 | 67 | ||
| NM_139612.3 | NP_647869.3 | 3 - 4 | 890 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001259684.1, NM_001259685.1 transcripts.
Set Membership: |
|

CG43367
Genomic Map