Assay Details
| Gene Symbol: CG43367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202119.2 | NP_001189048.2 | 18 - 19 | 6406 | 80 | |
| NM_001259684.1 | NP_001246613.1 | 17 - 18 | 6608 | 80 | ||
| NM_001259685.1 | NP_001246614.1 | 16 - 17 | 6255 | 80 | ||
| NM_139612.3 | NP_647869.3 | 17 - 18 | 6280 | 80 | ||
More Information
Set Membership: |
|

CG43367
Genomic Map