Assay Details
| Gene Symbol: CG43367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202119.2 | NP_001189048.2 | 21 - 22 | 6991 | 99 | |
| NM_001259684.1 | NP_001246613.1 | 20 - 21 | 7193 | 99 | ||
| NM_001259685.1 | NP_001246614.1 | 19 - 20 | 6840 | 99 | ||
| NM_139612.3 | NP_647869.3 | 20 - 21 | 6865 | 99 | ||
More Information
Set Membership: |
|

CG43367
Genomic Map