Assay Details
| Gene Symbol: CG43367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202119.2 | NP_001189048.2 | 22 - 23 | 7281 | 72 | |
| NM_001259684.1 | NP_001246613.1 | 21 - 22 | 7483 | 72 | ||
| NM_001259685.1 | NP_001246614.1 | 20 - 21 | 7130 | 72 | ||
| NM_139612.3 | NP_647869.3 | 21 - 22 | 7155 | 72 | ||
More Information
Set Membership: |
|

CG43367
Genomic Map