Assay Details
| Gene Symbol: CG43367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202119.2 | NP_001189048.2 | 24 - 25 | 8215 | 88 | |
| NM_001259684.1 | NP_001246613.1 | 23 - 24 | 8417 | 88 | ||
| NM_001259685.1 | NP_001246614.1 | 22 - 23 | 8064 | 88 | ||
| NM_139612.3 | NP_647869.3 | 23 - 24 | 8089 | 88 | ||
More Information
Set Membership: |
|

CG43367
Genomic Map