Assay Details
Gene Symbol: Ctl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274482.1 | NP_001261411.1 | 1 - 2 | 294 | 110 | |
NM_139627.2 | NP_647884.1 | 1 - 2 | 294 | 110 | ||
GenBank mRNA | BT150374.1 | - | 1 - 2 | 292 | 110 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
