Assay Details
| Gene Symbol: Ctl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139627.2 | NP_647884.1 | 5 - 6 | 1354 | 62 | |
| GenBank mRNA | BT150374.1 | - | 5 - 6 | 1352 | 62 | |
More Information
Set Membership: |
|

Ctl1
Genomic Map