Assay Details
| Gene Symbol: ATPsynCF6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139685.2 | NP_647942.1 | 1 - 2 | 333 | 120 | |
| GenBank mRNA | KX531574.1 | - | 1 - 2 | 295 | 120 | |
More Information
Set Membership: |
|

ATPsynCF6L
Genomic Map