Assay Details
| Gene Symbol: CHMP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139690.4 | NP_647947.1 | 1 | 756 | 104 | |
| GenBank mRNA | AY061433.1 | - | 1 | 702 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHMP2B| Gene Symbol: CHMP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139690.4 | NP_647947.1 | 1 | 756 | 104 | |
| GenBank mRNA | AY061433.1 | - | 1 | 702 | 104 | |
Set Membership: |
|