Assay Details
| Gene Symbol: Eaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139724.4 | NP_647981.1 | 2 - 3 | 433 | 76 | |
| GenBank mRNA | AY071012.1 | - | 2 - 3 | 374 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eaf6| Gene Symbol: Eaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139724.4 | NP_647981.1 | 2 - 3 | 433 | 76 | |
| GenBank mRNA | AY071012.1 | - | 2 - 3 | 374 | 76 | |
Set Membership: |
|