Assay Details
| Gene Symbol: lin-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139726.2 | NP_647983.1 | 3 - 4 | 654 | 70 | |
| GenBank mRNA | AF521096.2 | - | 3 - 4 | 651 | 70 | |
| AY118360.1 | - | 3 - 4 | 651 | 70 | ||
More Information
Set Membership: |
|

lin-28
Genomic Map