Assay Details
| Gene Symbol: Ppat-Dpck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139728.2 | NP_647985.1 | 1 - 2 | 598 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppat-Dpck| Gene Symbol: Ppat-Dpck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139728.2 | NP_647985.1 | 1 - 2 | 598 | 76 | |
Set Membership: |
|