Assay Details
| Gene Symbol: DNApol-epsilon58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139738.2 | NP_647995.1 | 1 - 2 | 768 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNApol-epsilon58| Gene Symbol: DNApol-epsilon58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139738.2 | NP_647995.1 | 1 - 2 | 768 | 88 | |
Set Membership: |
|